missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRMT3 Antibody (RAB-C372) [Janelia Fluor™ 585], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-20143JF585
This item is not returnable.
View return policy
Description
PRMT3 Monoclonal antibody specifically detects PRMT3 in Human samples. It is validated for ELISA,Immunoprecipitation,Western Blot,Mass Spectrometry,ChIP assay (ChIP),Immunocytochemistry/ Immunofluorescence
Specifications
| PRMT3 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| EC 2.1.1, EC 2.1.1.-, heterogeneous nuclear ribonucleoprotein methyltransferase-like 3, Heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 3, HMT1 hnRNP methyltransferase-like 3, HMT1 hnRNP methyltransferase-like 3 (S. cerevisiae), HRMT1L3, protein arginine methyltransferase 3, protein arginine N-methyltransferase 3 | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing PRMT3 protein under non-denaturing conditions. | |
| 0.1 mL | |
| Chromatin Research, Core ESC Like Genes, Stem Cell Markers | |
| 10196 | |
| Store at 4°C in the dark. | |
| IgG κ |
| ELISA, Immunoprecipitation, Western Blot, Mass Spectrometry, ChIP Assay, Immunocytochemistry/Immunofluorescence | |
| RAB-C372 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction