missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRMT4 Polyclonal Antibody

PRMT4 Polyclonal Antibody
Brand: Invitrogen PA586984
This item is not returnable.
View return policy
Description
PRMT4 Polyclonal Antibody for Western Blot
Specifications
| PRMT4 | |
| Polyclonal | |
| Unconjugated | |
| Carm1 | |
| Carm1; coactivator associated arginine methyltransferase 1; coactivator-associated arginine methyltransferase 1; coactivator-associated arginine methyltransferase 1 variant 5; histone-arginine methyltransferase CARM1; PRMT4; protein arginine methyltransferase; protein arginine N-methyltransferase 4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10498, 363026, 59035 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q4AE70, Q86X55, Q9WVG6 | |
| Carm1 | |
| Recombinant Protein of human CARM1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction