Learn More
PRMT5 Monoclonal Antibody (OTI3E6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA807620S
Encompass_Preferred
 View more versions of this product
Product Code. 17538253
  • € 242.00 / 30µL
Description
PRMT5 Monoclonal antibody specifically detects PRMT5 in Human samples. It is validated for Western Blot
Specifications
Specifications
PRMT5
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q53HL2
Mouse
Affinity chromatography
RUO
55143
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI3E6
Unconjugated
CDCA8
72 kDa ICln-binding protein; EC 2.1.1.125; histone-arginine N-methyltransferase PRMT5; HMT1 hnRNP methyltransferase-like 5; HRMT1L5; hypothetical protein; I79_013398; IBP72; Jak-binding protein 1; JBP1; PRMT5; protein arginine methyltransferase 5; protein arginine N-methyltransferase 5; Protein arginine N-methyltransferase 5, N-terminally processed; protein arginine N-methyltransferase 5-like; shk1 kinase-binding protein 1 homolog; Skb1; SKB1 homolog; SKB1Hs
Full length human recombinant protein of human CDCA8 produced in E.coli.