Learn More
PRMT5 Mouse anti-Human, Clone: OTI4G7, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF807462
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Arginine methylation is an irreversible post translational modification which has only recently been linked to protein activity. At least three types of PRMT enzymes have been identified in mammalian cells. These enzymes have been shown to have essential regulatory functions by methylation of key proteins in several fundamental areas. These protein include nuclear proteins, IL enhancer binding factor, nuclear factors, cell cycle proteins, signal transduction proteins, apoptosis proteins, and viral proteins. The mammalian PRMT family currently consists of 7 members that share two large domains of homology. Outside of these domains, epitopes were identified and antibodies against all 7 PRMT members have been developed.Specifications
| PRMT5 | |
| Monoclonal | |
| Unconjugated | |
| PRMT5 | |
| HRMT1L5, IBP72, JBP1, SKB1, SKB1Hs | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10419 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin) | |
| OTI4G7 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O14744 | |
| PRMT5 | |
| Human recombit protein fragment corresponding to amino acids 238-499 of human PRMT5 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.