missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRMT6 Antibody (SR1719), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22024-100ul
This item is not returnable.
View return policy
Description
PRMT6 Monoclonal antibody specifically detects PRMT6 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| PRMT6 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| SR1719 | |
| Western Blot 1:500-1:1000 | |
| EC 2.1.1.-, EC 2.1.1.125, FLJ10559, FLJ51477, Heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 6, Histone-arginine N-methyltransferase PRMT6, HMT1 hnRNP methyltransferase-like 6, HMT1 hnRNP methyltransferase-like 6 (S. cerevisiae), HRMT1L6, protein arginine methyltransferase 6, protein arginine N-methyltransferase 6 | |
| A synthesized peptide derived from human PRMT6 (Uniprot #: Q96LA8) | |
| 100 μg | |
| Chromatin Research, Signal Transduction | |
| 55170 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction