missing translation for 'onlineSavingsMsg'
Learn More

PRMT6 Polyclonal Antibody
GREENER_CHOICE

PRMT6 Polyclonal Antibody

Brand:  Invitrogen PA580942

Product Code. 16348014

  • € 348.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PRMT6 Polyclonal Antibody for ICC/IF

TRUSTED_SUSTAINABILITY
Specifications

Specifications

PRMT6
Polyclonal
Unconjugated
PRMT6
AW124876; BB233495; FLJ10559; FLJ51477; heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 6; Histone-arginine N-methyltransferase PRMT6; HMT1 hnRNP methyltransferase-like 6; Hrmt1l6; PRMT6; PRMT-6; protein arginine methyltransferase 6; protein arginine N-methyltransferase 6
Rabbit
Antigen Affinity Chromatography, Protein A
RUO
55170
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
1 mg/mL
PBS with no preservative
Q96LA8
PRMT6
A synthetic peptide corresponding to the N-terminus of the Human PRMT6.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.