missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRMT7 Monoclonal Antibody (OTI10A8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809927S
This item is not returnable.
View return policy
Description
PRMT7 Monoclonal antibody specifically detects PRMT7 in Human samples. It is validated for Western BlotSpecifications
| PRMT7 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UQF2 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9479 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI10A8 | |
| Unconjugated | |
| MAPK8IP1 | |
| [Myelin basic protein]-arginine N-methyltransferase PRMT7; 4933402B05Rik; BC006705; FLJ10640; Histone-arginine N-methyltransferase PRMT7; KIAA1933; myelin basic protein-arginine N-methyltransferase; PRMT7; PRMT-7; protein arginine methyltransferase 7; protein arginine N-methyltransferase 7; RGD1304869 | |
| Full length human recombinant protein of human MAPK8IP1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction