missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRMT7 Monoclonal Antibody (OTI10A8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809927
This item is not returnable.
View return policy
Description
Arginine methylation is an irreversible post translational modification which has been implicated in RNA processing and trafficking, receptor-mediated signaling, and transcription. At least three types of PRMT enzymes have been identified in mammalian cells. These enzymes have been shown to have essential regulatory functions by methylation of key proteins in several fundamental areas. These proteins include nuclear proteins, IL enhancer binding factor, nuclear factors, cell cycle proteins, signal transduction proteins, apoptosis proteins, and viral proteins. The mammalian PRMT family currently consists of 7 members that share two large domains of homology. Outside of these domains, epitopes were identified and antibodies against all 7 PRMT members have been developed.Specifications
| PRMT7 | |
| Monoclonal | |
| Unconjugated | |
| PRMT7 | |
| SBIDDS | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 54496 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI10A8 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NVM4 | |
| PRMT7 | |
| Full length human recombit protein of human PRMT7 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction