missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ProANP Monoclonal Antibody (C6), Invitrogen™

Product Code. 17959882
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17959882 200 μL 200µL
17999682 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17959882 Supplier Invitrogen™ Supplier No. MA551085

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Receptor for the natriuretic peptide hormones, binding with similar affinities atrial natriuretic peptide NPPA/ANP, brain natriuretic peptide NPPB/BNP, and C-type natriuretic peptide NPPC/CNP. May function as a clearance receptor for NPPA, NPPB and NPPC, regulating their local concentrations and effects. May regulate diuresis, blood pressure and skeletal development. Does not have guanylate cyclase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ProANP
Applications Western Blot, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone C6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene NPPA
Gene Accession No. P01160, P01161
Gene Alias ANPRC; Atrial natriuretic peptide receptor 3; NPR3
Gene Symbols NPPA
Host Species Mouse
Immunogen Recombinant Human NT-ProANP protein, Ala25-Arg123 (Accession #P01160).
Purification Method Protein A/G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24602, 4878
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.