missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Procainamide Monoclonal Antibody (PC12-424.5)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA110760
This item is not returnable.
View return policy
Description
MA1-10760 detects Procainamide. MA1-10760 has been successfully used in ELISA applications. The MA1-10760 immunogen is Procainamide, conjugated to KLH.
Procainamide is an oral antiarrhythmic agent that has been in wide use for more than 60 years. Long term procainamide therapy is known to induce hypersensitivity reactions, autoantibody formation and a lupus-like syndrome but is a rare cause of clinically apparent acute liver injury. [National Center for Biotechnology Information. PubChem Compound Database; CID=4913].
Specifications
| Procainamide | |
| Monoclonal | |
| 0.8 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Mouse | |
| purified | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG2b |
| ELISA | |
| PC12-424.5 | |
| Unconjugated | |
| Biocoryl; C13H21N3O; Novocainamide; Novocamid; Procaine amide; Procan; Procanbid; Pronestyl | |
| Procainamide, conjugated to KLH. | |
| 500 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction