missing translation for 'onlineSavingsMsg'
Learn More

Procaine Hydrochloride, TRC

Product Code. 30133763
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133763 50 g 50g
30136731 10 g 10g
18080750 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133763 Supplier Toronto Research Chemicals Supplier No. TRCP75515050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-05-8
Formula Weight 272.1292
InChI Formula InChI=1S/C13H20N2O2.ClH/c1-3-15(4-2)9-10-17-13(16)11-5-7-12(14)8-6-11;/h5-8H,3-4,9-10,14H2,1-2H3;1H
IUPAC Name 2-(diethylamino)ethyl 4-aminobenzoate;hydrochloride
Molecular Formula C13 H20 N2 O2 . Cl H
Molecular Weight (g/mol) 272.77
SMILES Cl.CCN(CC)CCOC(=O)c1ccc(N)cc1
Synonym Benzoic acid, 4-amino-, 2-(diethylamino)ethyl ester, hydrochloride (1:1), Benzoic acid, 4-amino-, 2-(diethylamino)ethyl ester, monohydrochloride (9CI), Benzoic acid, p-amino-, 2-(diethylamino)ethyl ester, monohydrochloride (8CI), 2-(Diethylamino)ethyl 4-aminobenzoate hydrochloride, 2-Diethylaminoethyl p-aminobenzoate hydrochloride, Allocaine, Aminocaine, Anadolor, Anesthesol, Anestil, Atoxicocaine, Bernacaine, Cetain, Chlorocaine, Diethylaminoethanol 4-aminobenzoate hydrochloride, Enpro, Ethocain, Ethocaine, Eugerase, Geriocaine, Gero, Gerovital, Gerovital H3, Herocaine, Irocaine, Isocain, Isocaine, Isocaine-Heisler, Jenacaine, Juvocaine, Kerocaine, Lactocaine, Medaject, Naucain, Naucaine, Neocaine, Neotonocaine, Novocain, Novocaine, Novocaine hydrochloride, Omnicain, Paracain, Planocaine, Polocaine, Procaine hydrochloride, Procaine monohydrochloride, Rocain, Scurocaine, Sevicaine, Syncaine, Syntocain, Topokain, Westocaine
Chemical Name or Material Procaine hydrochloride

Specifications

Quantity 50 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.