missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Procalcitonin Antibody (9D9.E11.C3.E8.F4.D4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-44103
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, ELISA
Procalcitonin Monoclonal specifically detects Procalcitonin in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Procalcitonin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000-1:2000, ELISA 1:100-1:2000 | |
| P06881 | |
| CALCA | |
| Procalcitonin Antibody (9D9.E11.C3.E8.F4.D4) was produced in mice prepared by repeated immunizations with full-length recombinant human Procalcitonin protein. (Uniprot: P06881) | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 9D9.E11.C3.E8.F4.D4 | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| ATP2B2plasma membrane calcium ATPase, ATPase, Ca++ transporting, plasma membrane 4, DKFZp686G08106, DKFZp686M088, EC 3.6.3, EC 3.6.3.8, Matrix-remodeling-associated protein 1, matrix-remodelling associated 1, MXRA1, Plasma membrane calcium ATPase isoform 4, plasma membrane calcium pump, Plasma membrane calcium pump isoform 4, plasma membrane calcium-transporting ATPase 4, PMCA4b, PMCA4PMCA4x, sarcolemmal calcium pump | |
| Mouse | |
| 13.9 kDa | |
| 100 μg | |
| Cancer | |
| 796 | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. | |
| IgG1 λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction