Learn More
Miele Professional™ ProCare Lab 10 AT
Product Code. 32237206 Shop All Miele Products
Change view
Click to view available options
Quantity:
5 L
10 L
Unit Size:
10 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32237206 10 L 10 L
32237203 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 32237206 Supplier Miele Professional™ Supplier No. 12959980
  • ONLINE EXCLUSIVE PRICE
    € 135.38
    € 88.00 / 10 L
    Save € 47.38
    34% Off
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For intensive machine cleaning of laboratory glassware and utensils.
  • Alkaline cleaning agent containing surfactants
  • Free from phosphates, oxidants and silicates
  • Specially developed for Miele Professional washer-disinfectors
  • Suitable for stainless steel, laboratory glassware, ceramics and plastics
  • Good cleaning for oils, grease, resins and organic colourants
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleaning Agent
Color-coded Yes
Form Liquid
Scent Unscented
Sterility Non-sterile
Quantity 10 L