missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Procaterol hydrochloride
GREENER_CHOICE

Product Code. 18792604
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18792604

Brand: Tocris Bioscience™ 1102/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent β2 agonist

Specific and very potent β2 agonist.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62929-91-3
Molecular Formula C16H23ClN2O3

Specifications

Quantity 50 mg
Formula Weight 326.82
Purity >99%
Molecular Weight (g/mol) 326.821
Chemical Name or Material Procaterol hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.