missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Procollagen I N-Peptide Monoclonal Antibody (C6)
GREENER_CHOICE

Mouse Monoclonal Antibody

€ 208.00 - € 538.00

Specifications

Antigen Procollagen I N-Peptide
Clone C6
Concentration 1 mg/mL
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Applications Immunohistochemistry (Paraffin), Western Blot
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17878692
GREENER_CHOICE
View Documents
Invitrogen™
MA541662
200 μL
€ 538.00
200µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17674336
View Documents
Invitrogen™
MA543883
20 μL
€ 208.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Procollagen I N-Peptide Monoclonal Antibody for Western Blot, IHC (P)

Procollagen type I N-terminal propeptide (P1NP) is a specific marker of bone formation, reflecting the synthesis of type I collagen, which constitutes the major part of the bone matrix. P1NP is derived during the conversion of procollagen to collagen, produced by osteoblasts during bone formation. Its primary clinical application lies in the diagnosis and monitoring of metabolic bone diseases, particularly osteoporosis. In osteoporotic patients, P1NP levels can predict bone mineral density (BMD) responses to treatments like teriparatide and romosozumab. Low serum levels of P1NP are indicative of a high risk of BMD loss, providing a valuable tool for premenopausal women with systemic lupus erythematosus (SLE) and other conditions affecting bone health.
Specifications

Specifications

Procollagen I N-Peptide
1 mg/mL
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Mouse
RUO
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
P02452, P02454
1277, 29393
Recombinant protein Procollagen I N-Terminal Propeptide.
Antibody
C6
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Monoclonal
Liquid
IgG1 κ
Human, Rat
COL1A1
P1NP; Procollagen I N
COL1A1
Primary
Protein A/G
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.