Learn More
PROCR Monoclonal Antibody (OTI3E7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805524
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a receptor for activated protein C, a serine protease activated by and involved in the blood coagulation pathway. The encoded protein is an N-glycosylated type I membrane protein that enhances the activation of protein C. Mutations in this gene have been associated with venous thromboembolism and myocardial infarction, as well as with late fetal loss during pregcy.Specifications
| PROCR | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UNN8 | |
| PROCR | |
| Human recombit protein fragment corresponding to amino acids 1-210 of human PROCR produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3E7 | |
| Unconjugated | |
| PROCR | |
| CCCA, CCD41, EPCR | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10544 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |