Learn More
Procyanidin B1, MedChemExpress
Product Code. 16492549
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Product Code. 16492549
Brand: Medchem Express HYN0795
MedChemExpress Procyanidin B1 is a polyphenolic flavonoid isolated from commonly eaten fruits, binds to TLR4/MD-2 complex, and has anti-inflammatory activity.
Procyanidin B1 is a polyphenolic flavonoid isolated from commonly eaten fruits, binds to TLR4/MD-2 complex, and has anti-inflammatory activity.
Specifications
Chemical Name or Material Procyanidin B1
CAS 20315-25-7
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C30H26O12
Quantity 1 mg
Solubility Information DMSO : 25 mg/mL (43.21 mM; Need ultrasonic)
SMILES O[C@H]1[C@@H](C2=CC=C(O)C(O)=C2)OC3=CC(O)=CC(O)=C3[C@@H]1C4=C5C(C[C@H](O)[C@@H](C6=CC=C(O)C(O)=C6)O5)=C(O)C=C4O
Molecular Weight (g/mol) 578.52
Formula Weight 578.52
Percent Purity 99.28%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only