missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Procyanidin B2, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | Procyanidin B2 |
| CAS | 29106-49-8 |
| Molecular Formula | C30 H26 O12 |
| Quantity | 5 mg |
| Synonym | [4,8'-Bi-2H-1-benzopyran]-3,3',5,5',7,7'-hexol','2,2'-bis(3,4-dihydroxyphenyl)-3,3',4,4'-tetrahydro-','(2R,2'R,3R,3'R,4R)-, [4,8'-Bi-2H-1-benzopyran]-3,3',5,5',7,7'-hexol','2,2'-bis(3,4-dihydroxyphenyl)-3,3',4,4'-tetrahydro-','[2R-[2α,3α,4β(2'R*,3'R*)]]-, [4,8''-Biflavan]-3,3',3'',3''',4',4''',5,5'',7,7''-decol','stereoisomer (8CI), (2R,2'R,3R,3'R,4R)-2,2'-Bis(3,4-dihydroxyphenyl)-3,3',4,4'-tetrahydro[4,8'-bi-2H-1-benzopyran]-3,3',5,5',7,7'-hexol, (+)-Procyanidin B2, (-)-Epicatechin-(4β→8)-(-)-epicatechin, Proanthocyanidin B2, Procyanidin B2, Procyanidol B2 |
| InChI Formula | InChI=1S/C30H26O12/c31-13-7-20(37)24-23(8-13)41-29(12-2-4-16(33)19(36)6-12)27(40)26(24)25-21(38)10-17(34)14-9-22(39)28(42-30(14)25)11-1-3-15(32)18(35)5-11/h1-8,10,22,26-29,31-40H,9H2/t22-,26-,27-,28-,29-/m1/s1 |
| SMILES | O[C@@H]1Cc2c(O)cc(O)c([C@@H]3[C@@H](O)[C@H](Oc4cc(O)cc(O)c34)c5ccc(O)c(O)c5)c2O[C@@H]1c6ccc(O)c(O)c6 |
| IUPAC Name | (2R,3R,4R)-2-(3,4-dihydroxyphenyl)-4-[(2R,3R)-2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxy-3,4-dihydro-2H-chromen-8-yl]-3,4-dihydro-2H-chromene-3,5,7-triol |
| Molecular Weight (g/mol) | 578.52 |
| Formula Weight | 578.1424 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction