missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: GNLGAMLRCV DLSRGLLEPP SLAEASLMQL KVTALTSTRL CKELASWVRR PGASLELSPE RLAEAMDSGQ NLQVSCLNAE Q Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 65%.
PRODH2 is similar to proline dehydrogenase (oxidase) 1, a mitochondrial enzyme which catalyzes the first step in proline catabolism. The function of this protein has not been determined.The protein encoded by this gene is similar to proline dehydrogenase (oxidase) 1, a mitochondrial enzyme which catalyzes the first step in proline catabolism. The function of this protein has not been determined.
Specifications
Specifications
| Antigen | PRODH2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | PRODH2 |
| Gene Accession No. | Q9UF12 |
| Gene Alias | 2510028N04Rik; 2510038B11Rik; HSPOX1; Hydroxyproline dehydrogenase; HYPDH; Kidney and liver proline oxidase 1; MmPOX; MmPOX1; PO; POX1; Probable proline dehydrogenase 2; probable proline oxidase 2; PRODH2; proline dehydrogenase (oxidase) 2; proline dehydrogenase 2; proline oxidase 1; proline oxidase-like protein |
| Gene Symbols | PRODH2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?