missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Profilin 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-23341
This item is not returnable.
View return policy
Description
Profilin 1 Polyclonal antibody specifically detects Profilin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications
| Profilin 1 | |
| Polyclonal | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100 | |
| profilin 1, Profilin I, profilin-1 | |
| Profilin synthetic peptide (coupled to carrier protein) that includes amino acids from the C-terminal region of human Profilin 1. This sequence is well conserved in rat and mouse Profilin 1, has some homology to Profilin-2a and Profilin-2b, and low homology to Profiin-3 and Profilin-4. [UniProt# P07737] | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Tumor Suppressors | |
| 5216 | |
| Store at -20°C. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS, 1 mg/ml BSA and 50% glycerol | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction