missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Profilin 1 Antibody (OTI1D5), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73605APC
This item is not returnable.
View return policy
Description
Profilin 1 Monoclonal specifically detects Profilin 1 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry.
Specifications
| Profilin 1 | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| PFN1 | |
| Full length recombinant protein of human Profilin 1 (NP_005013) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG2a |
| Flow Cytometry | |
| OTI1D5 | |
| Flow Cytometry | |
| profilin 1, Profilin I, profilin-1 | |
| Mouse | |
| Affinity Purified | |
| Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Tumor Suppressors | |
| 5216.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction