missing translation for 'onlineSavingsMsg'
Learn More

Profilin 2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 227.00 - € 326.00

Specifications

Antigen Profilin 2
Dilution Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:2500-1:10000
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18689517
View Documents
Novus Biologicals
NBP3-00301-100ul
100 μg
€ 326.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18647487
View Documents
Novus Biologicals
NBP3-00301-50ul
50 μg
€ 227.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Profilin 2 Polyclonal antibody specifically detects Profilin 2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin)
Specifications

Specifications

Profilin 2
Western Blot, ELISA, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cell Biology, Cellular Markers, Signal Transduction, Stem Cell Markers
0.2 um filtered solution in PBS
5217
IgG
Antigen and protein A Affinity-purified
Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:2500-1:10000
Polyclonal
Purified
RUO
Human
D3S1319E, PFL, profilin 2, Profilin II, profilin-2
Produced in rabbits immunized with purified, recombinant Human Profilin 2 (Accession#: AAH18049.1; Met1-Phe140)
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.