missing translation for 'onlineSavingsMsg'
Learn More

Profluralin, TRC

Product Code. 30412835
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30412835 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30412835 Supplier Toronto Research Chemicals Supplier No. TRCP7558935G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Profluralin
CAS 26399-36-0
Molecular Formula C14 H16 F3 N3 O4
Quantity 5 g
Synonym p-Toluidine','N-(cyclopropylmethyl)-α,α,α-trifluoro-2,6-dinitro-N-propyl- (8CI), N-(Cyclopropylmethyl)-2,6-dinitro-N-propyl-4-(trifluoromethyl)benzenamine, B 4576, CG 10832, CGA 10832, ER 5461, N-Cyclopropylmethyl-N-propyl-4-trifluoromethyl-2,6-dinitroaniline, Pregard, Profluralin, SGA 10832, Tolban
InChI Formula InChI=1S/C14H16F3N3O4/c1-2-5-18(8-9-3-4-9)13-11(19(21)22)6-10(14(15,16)17)7-12(13)20(23)24/h6-7,9H,2-5,8H2,1H3
SMILES CCCN(CC1CC1)c2c(cc(cc2[N+](=O)[O-])C(F)(F)F)[N+](=O)[O-]
IUPAC Name N-(cyclopropylmethyl)-2,6-dinitro-N-propyl-4-(trifluoromethyl)aniline
Molecular Weight (g/mol) 347.29
Formula Weight 347.1093
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.