Learn More
Progesterone Antibody (6-5E-3F), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-78056AF750
Product Code. 18272956
  • € 635.00 / 0.10mL
Description
Progesterone Monoclonal specifically detects Progesterone in All Species samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Progesterone
Monoclonal
Alexa Fluor 750
50mM Sodium Borate with 0.05% Sodium Azide
Mouse
Protein A or G purified
RUO
This monoclonal antibody is specific for progesterone. It exhibits minimal cross reactivity with related compounds in ELISA. It reacts with Progesterone-11a-HMS-BSA: 100%; 5-beta-Pregnane-3,20-dione: 48%; 5-alpha-Pregnane-3,20-dione: 26.4%; 17-alpha-Hydroxyprogesterone: 2.5%; 20-alpha-Hydroxyprogesterone: 0.04%. Progesterone is a steroid hormone synthesized from the cholesterol derivative, pregnenolone, in the cortex of the adrenal gland. Progesterone is secreted by the corpus luteum and acts to prepare the endometrium for the implantation of a fertilized egg. During pregnancy, it is secreted by the placenta to prevent spontaneous abortion and to stimulate the development of mammary tissue to produce milk. Thus, progesterone plays a central role in the reproductive events associated with the establishment and maintenance of pregnancy. Luteinized theca cells of normal ovary secrete progesterone. The determination of progesterone concentrations in the body fluids is of great value for endocrinological investigations in women. This monoclonal antibody may prove useful in identification of ovarian tumors.
Store at 4°C in the dark.
ELISA
6-5E-3F
ELISA
4-Pregnen-3,20-dione
Progesterone hemisuccinate conjugated to bovine serum albumin
0.1 mL
Primary
All species
IgG1 κ