missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Progesterone R/NR3C3 Antibody (Alpha PR6), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-2765JF646
This item is not returnable.
View return policy
Description
Progesterone R/NR3C3 Monoclonal specifically detects Progesterone R/NR3C3 in Human, Mouse, Rat, Bovine, Chicken, Guinea Pig, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| ProgesteroneR/NR3C3 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PGR | |
| Progesterone receptor purified from chick oviduct cytosol. | |
| 0.1 mL | |
| Primary | |
| Detects the B form of the progesterone receptor (PR). This does not cross-react with estrogen receptor or glucocorticoid receptor. | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Alpha PR6 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| NR3C3PRNuclear receptor subfamily 3 group C member 3, progesterone receptor | |
| Mouse | |
| Protein A purified | |
| Breast Cancer, Cancer | |
| 5241 | |
| Bovine, Chicken, Guinea Pig, Human, Mouse, Primate, Rabbit, Rat, Sheep | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction