missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Progesterone R/NR3C3 Antibody (OTI11E8), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-73607CL1
This item is not returnable.
View return policy
Description
Progesterone R/NR3C3 Monoclonal antibody specifically detects Progesterone R/NR3C3 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| Progesterone R/NR3C3 | |
| Monoclonal | |
| CoraFluor 1 | |
| NR3C3PRNuclear receptor subfamily 3 group C member 3, progesterone receptor | |
| Human recombinant protein fragment corresponding to amino acids 1-298 of human PGR (NP_000917) produced in E.coli. | |
| 0.1 mL | |
| Breast Cancer, Cancer, Immunology, Innate Immunity, Phospho Specific, Signal Transduction | |
| 5241 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| OTI11E8 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction