missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Progesterone R/NR3C3 Antibody (OTI11E8), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73607C
This item is not returnable.
View return policy
Description
Progesterone R/NR3C3 Monoclonal specifically detects Progesterone R/NR3C3 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Progesterone R/NR3C3 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PGR | |
| Recombinant protein fragment corresponding to amino acids 1-298 of human Progesterone R/NR3C3 (NP_000917) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI11E8 | |
| Western Blot, Immunohistochemistry | |
| NR3C3PRNuclear receptor subfamily 3 group C member 3, progesterone receptor | |
| Mouse | |
| Affinity Purified | |
| Breast Cancer, Cancer, Phospho Specific | |
| 5241.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction