missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Progesterone R/NR3C3 Antibody (PR484), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-33325DL594
This item is not returnable.
View return policy
Description
Progesterone R/NR3C3 Monoclonal antibody specifically detects Progesterone R/NR3C3 in Human samples. It is validated for Simple Western, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
Specifications
| Progesterone R/NR3C3 | |
| Monoclonal | |
| DyLight 594 | |
| NR3C3PRNuclear receptor subfamily 3 group C member 3, progesterone receptor | |
| Recombinant full-length human Progesterone R/NR3C3 protein (PGR) (Uniprot: P06401 ) | |
| 0.1 mL | |
| Breast Cancer, Cancer | |
| 5241 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Simple Western, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| PR484 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction