missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Progesterone R/NR3C3 Antibody (SPM566), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34429AF594
This item is not returnable.
View return policy
Description
Progesterone R/NR3C3 Monoclonal specifically detects Progesterone R/NR3C3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Progesterone R/NR3C3 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PGR | |
| Recombinant full-length human Progesterone R/NR3C3 protein (PGR) (Uniprot: P06401 ) | |
| 0.1 mL | |
| Breast Cancer, Cancer, Phospho Specific | |
| This monoclonal antibody is specific to progesterone receptor and shows minimal cross-reaction with other members of the family. Progesterone receptor is expressed as two major isoforms, PR-A (81kDa) and PR-B (116kDa). Expression of PgR has been suggested to reflect a intact estrogen regulatory machinery and therefore, predict better clinical response to endocrine therapy than ER alone. It is excellent for immunohistochemical staining of formalin/paraffin tissues. | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| SPM566 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| NR3C3PRNuclear receptor subfamily 3 group C member 3, progesterone receptor | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction