missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Progesterone Receptor Ab-8 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ Progesterone Receptor Ab-8, Mouse Monoclonal Antibody.
€ 84.70 - € 710.00
Specifications
Antigen | Progesterone Receptor Ab-8 |
---|---|
Clone | hPRa 2+hPRa 3 |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12680867
|
Epredia
MS-298-P1ABX |
Concentrated | 100 μL |
€ 627.00
100µL |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12650867
|
Epredia
MS-298-P |
Concentrated | 1 mL |
€ 594.00
1mL |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12690867
|
Epredia
MS-298-PABX |
Concentrated | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12660867
|
Epredia
MS-298-P0 |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12630877
|
Epredia
MS-298-R7 |
Prediluted | 7 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12640877
|
Epredia
MS-298-RQ |
Ready-to-Use | 12 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Human PgR exists in two forms: 116kDa (B-form) and 81kDa (A-form). It acts as ligand activated transcription factor to regulate expression of the target genes. Null mutation in PgR gene leads to pleiotrophic reproductive abnormalities.Host Species: Mouse
Clone: hPRa 2 + hPRa 3
Isotype: IgG1/κ + IgG1/κ
Species Reactivity: Human, Horse, Sheep and Pig. Others not known.
Epitope: N-terminal half of human PgR
Immunogen: PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice
Molecular Weight: 116kDa (B-form) and 81kDa (A-form)
Positive Control: T47D or Breast CA
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native and denatured)
- Sedimentation in Sucrose Gradient
Specifications
Progesterone Receptor Ab-8 | |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
ASR, IVD | |
PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice |
hPRa 2+hPRa 3 | |
Monoclonal | |
Mouse | |
Cancer and Tumor Biology | |
Primary |