missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prohibitin Antibody (PHB/3230), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08687AF647
This item is not returnable.
View return policy
Description
Prohibitin Monoclonal antibody specifically detects Prohibitin in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin), Protein Array
Specifications
| Prohibitin | |
| Monoclonal | |
| Alexa Fluor 647 | |
| PHB1, prohibitin | |
| Recombinant human Prohibitin protein fragment (aa167-261) (exact sequence is proprietary) (Uniprot: P35232) | |
| 100 μg | |
| Cell Cycle and Replication, Core ESC Like Genes, Ovarian Carcinoma Cell Markers, Stem Cell Markers | |
| 5245 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, Immunohistochemistry (Paraffin), Protein Array | |
| PHB/3230 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction