missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prohibitin, Mouse anti-Human, Clone: PHB/3225, Novus Biologicals™
Mouse Monoclonal Antibody
€ 281.00 - € 512.00
Specifications
| Antigen | Prohibitin |
|---|---|
| Clone | PHB/3225 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry 1-2 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18252067
|
Novus Biologicals
NBP2-79748-20UG |
20 μg |
€ 281.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18224228
|
Novus Biologicals
NBP2-79748-100UG |
100 μg |
€ 512.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Prohibitin Monoclonal specifically detects Prohibitin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Protein Array.Specifications
| Prohibitin | |
| 0.2 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Cell Cycle and Replication, Core ESC Like Genes, Ovarian Carcinoma Cell Markers, Stem Cell Markers | |
| 10 mM PBS with 0.05% BSA | |
| PHB1, prohibitin | |
| PHB | |
| IgG2b κ | |
| Protein A or G purified | |
| Recognizes a protein of 30kDa which is identified as Prohibitin, an evolutionarily conserved protein with homologues found in yeast to man and located in the inner membrane of mitochondria. |
| PHB/3225 | |
| Western Blot 1-2 ug/ml, Immunohistochemistry 1-2 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P35232, P35232 | |
| 5245 | |
| Recombinant human Prohibitin protein fragment (aa167-261) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title