missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Prohibitin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140989
This item is not returnable.
View return policy
Description
Prohibitin Polyclonal Antibody for Western Blot, IHC (P)
Prohibitin is from an evolutionarily conserved gene that is ubiquitously expressed. It is thought to be a negative regulator of cell proliferation and may be a tumor suppressor. Mutations in PHB have been linked to sporadic breast cancer. Prohibitin is expressed as two transcripts with varying lengths of 3' untranslated region. The longer transcript is present at higher levels in proliferating tissues and cells, suggesting that this longer 3' untranslated region may function as a trans-acting regulatory RNA.
Specifications
| Prohibitin | |
| Polyclonal | |
| Unconjugated | |
| PHB | |
| BAP 32; Bap32; B-cell receptor-associated protein 32; epididymis luminal protein 215; epididymis secretory sperm binding protein Li 54e; fa18f07; fb09f05; HEL-215; HEL-S-54e; OTTHUMP00000165765; OTTHUMP00000165766; OTTHUMP00000220103; OTTHUMP00000220170; OTTHUMP00000220171; PHB; phb protein; PHB1; Prohibitin; sr:nyz128; wu:fa18f07; wu:fb09f05 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18673, 25344, 5245 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P35232, P67778, P67779 | |
| PHB | |
| Synthetic peptide within C-terminal human Prohibitin. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction