missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Prohibitin Polyclonal Antibody
GREENER_CHOICE

Product Code. 17908932
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17908932 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17908932 Supplier Invitrogen™ Supplier No. PA5144101

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Prohibitin Polyclonal Antibody for Western Blot, IHC (P), IP, ELISA

Prohibitin is from an evolutionarily conserved gene that is ubiquitously expressed. It is thought to be a negative regulator of cell proliferation and may be a tumor suppressor. Mutations in PHB have been linked to sporadic breast cancer. Prohibitin is expressed as two transcripts with varying lengths of 3' untranslated region. The longer transcript is present at higher levels in proliferating tissues and cells, suggesting that this longer 3' untranslated region may function as a trans-acting regulatory RNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prohibitin
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 3.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene PHB
Gene Accession No. P35232
Gene Alias BAP 32; Bap32; B-cell receptor-associated protein 32; epididymis luminal protein 215; epididymis secretory sperm binding protein Li 54e; fa18f07; fb09f05; HEL-215; HEL-S-54e; OTTHUMP00000165765; OTTHUMP00000165766; OTTHUMP00000220103; OTTHUMP00000220170; OTTHUMP00000220171; PHB; phb protein; PHB1; Prohibitin; sr:nyz128; wu:fa18f07; wu:fb09f05
Gene Symbols PHB
Host Species Rabbit
Immunogen Recombinant Human Prohibitin protein (1-272AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5245
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.