Learn More
PROK2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA5142524
Additional Details : Weight : 0.01000kg
Beschreibung
Additional Information: This antibody is expected to recognize isoform a (NP_001119600.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000. Preliminary analysis showed no signal in Western Blot.
Prokineticin-2 (PK2) is a cysteine-rich secreted protein that is expressed in the suprachiasmatic nucleus (SCN) with receptors located in the critical autonomic control centers of the brain. It has a depolarizing effect on neurons expressing the receptor. PK2 is predominantly controlled by the endogenous circadian clock, but light also plays a modulatory role. PK2 functions as a critical SCN output molecule responsible for circadian locomotor rhythms. PK2 expression is high during the day, and responsive to nocturnal light pulses. PK2 also functions as a chemoattractant for subventricular zone-derived neuronal progenitors.Spezifikation
PROK2 | |
Polyclonal | |
Unconjugated | |
PROK2 | |
Bombina variegata 8 kD protein; Bombina variegata 8kDa protein; Bv 8 homolog (Bombina variegata); BV8; Bv8/prokineticin 2-like protein; Bv8/prokineticin 2-like protein splice variant; HH4; homolog of mouse Bv8 (Bombina variegata 8 kDa); homolog of mouse Bv8 (Bombina variegata 8 kDa); prokineticin 2; KAL4; MIT1; PK2; Prok1; Prok2; prokineticin 1; prokineticin 2; prokineticin 2 beta; prokineticin-2; Protein Bv8 homolog; secreted protein | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
60675 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG |
Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9HC23 | |
PROK2 | |
Peptide with sequence C-HPLTRKNNFGNGRQE. | |
100 μg | |
Primary | |
Human | |
Liquid |