missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prokineticin R1/PROKR1 Antibody (Multiple seq-one in animal), Alexa Fluor™ 350, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-27975AF350
This item is not returnable.
View return policy
Description
Prokineticin R1/PROKR1 Monoclonal antibody specifically detects Prokineticin R1/PROKR1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Prokineticin R1/PROKR1 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| G protein-coupled receptor 73, G protein-coupled receptor ZAQ, GPR73, GPR73aG-protein coupled receptor 73, PK-R1, PKR1G-protein coupled receptor ZAQ, prokineticin receptor 1, ZAQ | |
| GPR73/ PROKR1 | |
| 0.1 mL | |
| GPCR | |
| 10887 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Multiple seq-one in animal | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction