Learn More
Prolactin Antibody (CL6550) [mFluor Violet 450 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-76502MFV450
Product Code. 30290357
  • € 770.00 / 0.10mL
Description
Prolactin Monoclonal antibody specifically detects Prolactin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Prolactin
Monoclonal
mFluor Violet 450 SE
prolactin
This antibody was developed using a recombinant protein derived from P01236, with the exact immunogen sequence remaining proprietary.
0.1 mL
Biologically Active Proteins, Cancer, Phospho Specific, Signal Transduction
5617
Store at 4⪚C in the dark.
IgG2b
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL6550
50mM Sodium Borate
Mouse
Protein A purified