missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prolactin R Antibody (B6.2 + PRLR742), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34555AF488
This item is not returnable.
View return policy
Description
Prolactin R Monoclonal specifically detects Prolactin R in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Prolactin R | |
| Monoclonal | |
| Alexa Fluor 488 | |
| delta 4-delta 7/11 truncated prolactin receptor, delta 4-SF1b truncated prolactin receptor, hPRL receptor, hPRLrI, PRL-R, prolactin receptor, prolactin receptor delta 7/11, secreted prolactin binding protein | |
| Mouse | |
| 70 kDa | |
| 0.1 mL | |
| Apoptosis | |
| 5618 | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| B6.2 + PRLR742 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| PRLR | |
| Semi-Purified human Prolactin R (B6.2); Recombinant human Prolactin R (PRLR742) (Uniprot: P16471) | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| It recognizes a protein of 70kDa, identified as prolactin receptor. Prolactin is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. The initial step in its action is the binding to a specific membrane receptor (prolactin receptor), which belongs to the superfamily of class 1 cytokine receptors. The function of the prolactin receptor is mediated, at least in part, by two families of signaling molecules: Janus kinases and signal transducers and activators of transcription. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction