missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prolactin R Antibody (SR2303), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 359.00 - € 449.00
Specifications
| Antigen | Prolactin R |
|---|---|
| Clone | SR2303 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18604334
|
Novus Biologicals
NBP3-21817-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18641305
|
Novus Biologicals
NBP3-21817-50ul |
50 μg |
€ 359.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Prolactin R Monoclonal antibody specifically detects Prolactin R in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
| Prolactin R | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| delta 4-delta 7/11 truncated prolactin receptor, delta 4-SF1b truncated prolactin receptor, hPRL receptor, hPRLrI, PRL-R, prolactin receptor, prolactin receptor delta 7/11, secreted prolactin binding protein | |
| A synthesized peptide derived from human Prolactin R (Uniprot #: P16471) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR2303 | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Apoptosis | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 5618 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title