missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prolactin R Antibody (U5), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 13 publications
€ 213.00
Specifications
| Antigen | Prolactin R |
|---|---|
| Clone | U5 |
| Dilution | Western Blot 1 μg/mL, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Prolactin R Monoclonal specifically detects Prolactin R in Human, Mouse, Rat, Porcine, Bovine, Equine, Hamster, Rabbit, Sheep samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Prolactin R | |
| Western Blot 1 μg/mL, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| delta 4-delta 7/11 truncated prolactin receptor, delta 4-SF1b truncated prolactin receptor, hPRL receptor, hPRLrI, PRL-R, prolactin receptor, prolactin receptor delta 7/11, secreted prolactin binding protein | |
| PRLR | |
| IgG1 | |
| Protein G purified | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| U5 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Apoptosis | |
| P16471 | |
| 5618 | |
| Purified rat liver PRL receptor. | |
| Primary | |
| Reconstitute with 0.1 mL PBS. | |
| Not yet tested in other species. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title