Learn More
Invitrogen™ Prolgolimab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA559692
 Afficher plus de versions de ce produit
Code produit. 30282817
  • € 1204.00 / 1mg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
BCD-100 (Prolgolimab): This product is a recombinant human anti-PD-1 monoclonal antibody.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
Prolgolimab
Recombinant Monoclonal
Unconjugated
BCD-100
Protein A
RUO
Human
Antibody
IgG1
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized