missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Proliferin Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547169
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Specifications
| Proliferin | |
| Polyclonal | |
| Unconjugated | |
| Prl2c2 | |
| Mitogen-regulated protein 1; MRP; Mrp1; MRP-1; Plf; Plf1; PLF-1; Prl2c2; prolactin family 2, subfamily c, member 2; prolactin-2C2; proliferin; proliferin-1 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 18811 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P04095 | |
| Prl2c2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Proliferin Phe30-Cys224. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction