missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promethazine Sulfone Hydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
Promethazine Sulfone Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Promethazine Sulfone Hydrochloride |
| CAS | 15374-15-9 |
| Analyte or Component Names | Promethazine Sulfone Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H20 N2 O2 S . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | N,N,alpha-Trimethyl-10H-phenothiazine-10-ethanamine 5,5-Dioxide Hydrochloride (1:1), 10H-Phenothiazine-10-ethanamine, N,N,alpha-trimethyl-, 5,5-dioxide, monohydrochloride (9CI), Phenothiazine, 10-(2-dimethylaminopropyl)-, dioxide, hydrochloride (6CI), Phenothiazine, 10-[2-(dimethylamino)propyl]-, 5,5-dioxide, monohydrochloride (8CI), Dioxopromethazine hydrochloride, N-(2-Dimethylaminopropyl)phenothiazine S,S-dioxide hydrochloride |
| InChI Formula | InChI=1S/C17H20N2O2S.ClH/c1-13(18(2)3)12-19-14-8-4-6-10-16(14)22(20,21)17-11-7-5-9-15(17)19;/h4-11,13H,12H2,1-3H3;1H |
| SMILES | Cl.CC(CN1c2ccccc2S(=O)(=O)c3ccccc13)N(C)C |
| IUPAC Name | 1-(5,5-dioxophenothiazin-10-yl)-N,N-dimethylpropan-2-amine;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction