missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propantheline-d3 Bromide (N-methyl-d3), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Propantheline-D3 (N-methyl-D3) Bromide |
| CAS | 64717-35-7 |
| Molecular Formula | C23 D3 H27 N O3 . Br |
| Quantity | 0.005 g |
| Synonym | 2-Propanaminium, N-(methyl-d3)-N-(1-methylethyl)-N-[2-[(9 H-xanthen-9-ylcarbonyl)oxy]ethyl]-, bromide (9 CI), Propantheline-D3 (N-methyl-D3) bromide, Propantheline-d3 bromide (N-methyl-d3), N-(Methyl-d3)-N-(1-methylethyl)-N-[2-[(9 H-xanthen-9-ylcarbonyl)oxy]ethyl]-2-propanaminium bromide, N-(Methyl-d3)-N,N-bis(1-methylethyl)-2-[(9 H-xanthen-9-ylcarbonyl)oxy]ethanaminium bromide, (2-Hydroxyethyl)diisopropyl(methyl-d3)ammonium bromide xanthene-9-carboxylate, β-Diisopropylaminoethyl xanthene-9-carboxylate methobromide-d3 |
| InChI Formula | InChI=1 S/C23H30NO3.BrH/c1-16(2)24(5,17(3)4)14-15-26-23(25)22-18-10-6-8-12-20(18)27-21-13-9-7-11-19(21)22;/h6-13,16-17,22 H,14-15H2,1-5H3;1 H/q+1;/p-1/i5D3; |
| SMILES | [Br-].[2 H]C([2 H])([2 H])[N+](CCOC(=O)C1c2ccccc2Oc3ccccc13)(C(C)C)C(C)C |
| IUPAC Name | di(propan-2-yl)-(trideuteriomethyl)-[2-(9 H-xanthene-9-carbonyloxy)ethyl]azanium;bromide |
| Molecular Weight (g/mol) | 451.412 |
| Formula Weight | 450.16 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction