Learn More
Bochem™ Propeller Stirrers
Click to view available options
Length (Metric):
400 mm
Unit Size:
Each
Description
- Flexible blades
- Rpm-range middle
- Mixing of media with low and middle viscosity
- Radial flow
Specifications
Specifications
| Length (Metric) | 400 mm |
| Quantity | 1/Pk. |
| Type | Propeller Stirrers |
| Material | Stainless Steel (18/10) |