missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Properdin Antibody (Novelmed patent anti-Properdin) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28394-100ug
This item is not returnable.
View return policy
Description
Properdin Monoclonal antibody specifically detects Properdin in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Properdin | |
| Monoclonal | |
| Unconjugated | |
| P27918 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Novelmed patent anti-Properdin | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| BFD, Complement factor P, complement factor properdin, PFCcomplement, PFD, PROPERDIN | |
| Complement Factor P/ Properdin | |
| 100 μg | |
| Immunology | |
| 5199 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction