Learn More
Propionaldehyde 2,4-Dinitrophenylhydrazone-d3, TRC
Specifications
Specifications
| Chemical Name or Material | Propionaldehyde 2,4-Dinitrophenylhydrazone-d3 |
| CAS | 259824-53-8 |
| Molecular Formula | C9H7D3N4O4 |
| Quantity | 50 mg |
| Synonym | Propanal 2-(2,4-Dinitrophenyl)hydrazone-d3, Propanal (2,4-Dinitrophenyl)hydrazone-d3, Propionaldehyde (2,4-Dinitrophenyl)hydrazone-d3 |
| InChI Formula | InChI=1S/C9H10N4O4/c1-2-5-10-11-8-4-3-7(12(14)15)6-9(8)13(16)17/h3-6,11H,2H2,1H3/i3D,4D,6D |
| SMILES | [2H]C1=C([2H])C([N+]([O-])=O)=C([2H])C([N+]([O-])=O)=C1N/N=C/CC |
| IUPAC Name | 1-(2,4-dinitrophenyl-3,5,6-d3)-2-propylidenehydrazine |
| Molecular Weight (g/mol) | 241.22 |
| Formula Weight | 241.09 |