missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propionic anhydride, 97%
Click to view available options
Quantity:
100 g
250 g
500 g
1000 g
5000 g
Unit Size:
1000g
100g
250g
5000g
500g
Description
Propionic anhydride is used as a esterifying reagent and as a reagent in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -45°C |
| Boiling Point | 166°C to 168°C |
| Quantity | 250 g |
| Percent Purity | 97% |
| Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction