missing translation for 'onlineSavingsMsg'
Learn More

Propranolol hydrochloride, MedChemExpress

Product Code. 16442176
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16442176 500 mg 500mg
16408376 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16442176 Supplier Medchem Express Supplier No. HYB0573
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Propranolol hydrochloride is a nonselective β-adrenergic receptor (βAR) antagonist, has high affinity for the β1AR and β2AR with Ki values of 1.8 nM and 0.8 nM, respectively.

Propranolol hydrochloride inhibits [3H]-DHA binding to rat brain membrane preparation with an IC50 of 12 nM. Propranolol hydrochloride is used for study of hypertension, pheochromocytoma, myocardial infarction, cardiac arrhythmias, angina pectoris, and hypertrophic cardiomyopathy.

Specifications

Chemical Name or Material Propranolol hydrochloride
CAS 318-98-9
Color White
Purity Grade Notes Research
Molecular Formula C16H22ClNO2
Quantity 500 mg
Solubility Information DMSO : ≥ 150 mg/mL (507.10 mM) ∣H2O : 33.33 mg/mL (112.68 mM; Need ultrasonic)
SMILES OC(CNC(C)C)COC1=CC=CC2=CC=CC=C12.Cl
Molecular Weight (g/mol) 295.8
Formula Weight 295.8
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.