missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proprotein Convertase 7/PCSK7 Antibody (OTI1B8), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73621AF750
This item is not returnable.
View return policy
Description
Proprotein Convertase 7/PCSK7 Monoclonal specifically detects Proprotein Convertase 7/PCSK7 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| Proprotein Convertase 7/PCSK7 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| PCSK7 | |
| Full length recombinant protein of human Proprotein Convertase 7/PCSK7 (NP_004707) produced in HEK293T cells. | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI1B8 | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| EC 3.4.21, EC 3.4.21.-, EC 3.4.21.61, EC 3.4.21.75, EC 3.4.21.93, hPC8, LPCFLJ23503, Lymphoma proprotein convertase, PC7FLJ45329, PC8prohormone convertase PC7, Prohormone convertase 7, Proprotein convertase 7, Proprotein convertase 8, proprotein convertase subtilisin/kexin type 7, SPC7proprotein convertase PC7, Subtilisin/kexin-like protease PC7 | |
| Mouse | |
| 0.1 mL | |
| 9159 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction